| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010790.hg.1 |
ENST00000582929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010790.hg.1 |
NR_036208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021370.hg.1 |
NR_036208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021370.hg.1 |
TC19001566.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900189245.hg.1 |
ENST00000582929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189245.hg.1 |
NR_036208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189245.hg.1 |
TC1900010790.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189245.hg.1 |
ENST00000582929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189245.hg.1 |
NR_036208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010790.hg.1 |
ENST00000582929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010790.hg.1 |
NR_036208.1 |
Refseq |