Assay Details
| Gene Symbol: MIR4327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036221.1 | 1 | 39 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | keratin associated protein 13-2 | KRTAP13-2 | KAP13-2 | -3.1kb | Sense | Chr. 21 - 30371391 - 30372239 on NCBI Build GRCh38 |
| Downstream | keratin associated protein 13-1 | KRTAP13-1 |
KAP13.1, KRTAP13.1 |
20.7kb | Anti-sense | Chr. 21 - 30396074 - 30396822 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2100007890.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000370.hg.1
Set Membership: |
|

MIR4327
Genomic Map