| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006638.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19006638.hg.1 |
TC19000501.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159129.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159129.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159129.hg.1 |
TC1900007941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159129.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159129.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159131.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159131.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159131.hg.1 |
TC1900007941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159131.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159131.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007941.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159128.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159128.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159128.hg.1 |
TC1900007941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159128.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159128.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000525484.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000532599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
ENST00000534349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006798.hg.1 |
NR_073431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159130.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159130.hg.1 |
NR_036504.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900159130.hg.1 |
TC1900007941.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900159130.hg.1 |
ENST00000592087.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900159130.hg.1 |
NR_036504.1 |
Refseq |
| Human Genome U95 Set |
78027_PM_at |
- |
- |
| Human Genome U95 Set |
78027_at |
- |
- |