Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc and ring finger 2 pseudogene 2 | ZNRF2P2 | -173.0 | Anti-sense | Chr. 7 - 29684772 - 29685821 on NCBI Build GRCh38 | |
| Downstream | probable C-mannosyltransferase DPY19L2 | LOC101928208 | 33.1kb | Sense | Chr. 7 - 29720476 - 29742396 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map