Assay Details
| Gene Symbol: MIR3663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037436.1 | 1 | 47 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR3663 host gene | MIR3663HG | Sense | Chr. 10 - 117153521 - 117169055 on NCBI Build GRCh38 | ||
| Downstream | potassium two pore domain channel subfamily K member 18 | KCNK18 |
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK |
29.7kb | Anti-sense | Chr. 10 - 117197489 - 117210299 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1000011942.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001686.hg.1
Set Membership: |
|

MIR3663
Genomic Map