Assay Details
| Gene Symbol: LINC00616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037866.1 | 1 - 2 | 268 | 74 | ||
| GenBank mRNA | BC042886.1 | - | 1 - 2 | 268 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105377443 | LOC105377443 | -525.9kb | Anti-sense | Chr. 4 - 137545641 - 137603430 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 7 member 11 | SLC7A11 |
CCBR1, xCT |
33.4kb | Sense | Chr. 4 - 138164094 - 138242418 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001569.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021956.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021956.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021956.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021956.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021956.hg.1 | TC04001569.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021958.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021958.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021958.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021958.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021958.hg.1 | TC04001569.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181810.hg.1 | TC0400011919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181812.hg.1 | TC0400011919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000503208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000505874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000512536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000513895.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | ENST00000514600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011919.hg.1 | NR_037866.1 | Refseq |

LINC00616
Genomic Map