Assay Details
| Gene Symbol: LOC100507053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037884.1 | 1 - 2 | 433 | 65 | ||
| GenBank mRNA | BX647987.1 | - | 1 - 2 | 434 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
alcohol dehydrogenase 5 (class III), chi polypeptide |
ADH5 |
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p |
-69.0 | Anti-sense | Chr. 4 - 99070978 - 99088788 on NCBI Build GRCh38 |
| Downstream | proliferating cell nuclear antigen pseudogene 1 | PCNAP1 | p1PCNA | 26.8kb | Anti-sense | Chr. 4 - 99160593 - 99161647 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000514.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04007555.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007555.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007555.hg.1 | TC04000514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007558.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007558.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007558.hg.1 | TC04000514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007559.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007559.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007559.hg.1 | TC04000514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007554.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007554.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007554.hg.1 | TC04000514.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156615.hg.1 | TC0400008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156614.hg.1 | TC0400008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000499178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506160.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000506454.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000509939.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | ENST00000510764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008231.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156618.hg.1 | TC0400008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | ENST00000500358.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | NR_037884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400156619.hg.1 | TC0400008231.hg.1 | Clariom D TC |

LOC100507053
Genomic Map