Assay Details
| Gene Symbol: NSUN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037947.1 | 5 - 6 | 916 | 70 | ||
| GenBank mRNA | AK055456.1 | - | 5 - 6 | 846 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1018 | LINC01018 | SRHC | -31.6kb | Anti-sense | Chr. 5 - 6582136 - 6588500 on NCBI Build GRCh38 |
| Downstream | steroid 5 alpha-reductase 1 | SRD5A1 | S5AR 1 | 10.1kb | Anti-sense | Chr. 5 - 6633322 - 6669562 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK055456.1 transcripts.
Set Membership: |
|

NSUN2
Genomic Map