| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
237137_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009340.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009340.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009340.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009340.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009340.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009340.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014757.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014757.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014757.hg.1 |
TC01000938.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100009340.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009340.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009340.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009340.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009340.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009340.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166565.hg.1 |
TC0100009340.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
ENST00000458748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
ENST00000602755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
NR_003023.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166566.hg.1 |
TC0100009340.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
237137_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
237137_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
237137_at1 |
- |
- |
| Human Genome U133 Set |
237137_at |
- |
- |
| Human Genome U133 Set |
237137_PM_at |
- |
- |