Assay Details
| Gene Symbol: RBAKDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015343.2 | 1 | 52 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RBAK-RBAKDN readthrough | RBAK-RBAKDN | RBAK-LOC389458 | Sense | Chr. 7 - 5045821 - 5073223 on NCBI Build GRCh38 | |
| Downstream |
zinc finger protein 890, pseudogene |
ZNF890P | 49.1kb | Anti-sense | Chr. 7 - 5121310 - 5144546 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003318.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000779.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000779.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000779.hg.1 | TC07003318.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000778.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000778.hg.1 | TC07003318.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196275.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196275.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196275.hg.1 | TC0700013332.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196276.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196276.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196276.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196276.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196276.hg.1 | TC0700013332.hg.1 | Clariom D TC |

RBAKDN
Genomic Map