Assay Details
| Gene Symbol: MAP2K4P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029423.1 | 1 | 46 | 74 | ||
| GenBank mRNA | AK056535.1 | - | 1 | 46 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | caudal type homeobox 4 | CDX4 | -108.4kb | Anti-sense | Chr. X - 73447254 - 73454585 on NCBI Build GRCh38 | |
| Downstream | cysteine rich hydrophobic domain 1 | CHIC1 | BRX | 63.0 | Anti-sense | Chr. X - 73563045 - 73687109 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001154.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17358194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788600 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32505206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11788615 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 144686 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 68374 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 6762 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 427363 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 402633 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 621124 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 762053 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 34484 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 804194 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 101357 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 42418 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 551181 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 439957 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P60271144 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21495252 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03718048 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21495258 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21495252 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3653330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788645 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788645 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788641 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11788642 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32505261 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P21495252 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03718048 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P21495252 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 44852 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 151235 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 176849 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 34329 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P17200323 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21495252 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P21495254 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41702512 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03718048 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21495256 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21495252 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21495255 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015609.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015609.hg.1 | TC0X001154.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | ENST00000438453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015610.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015610.hg.1 | TC0X001154.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | ENST00000438453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | ENST00000438453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168640.hg.1 | TC0X00010056.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00168641.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168641.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168641.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168641.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168641.hg.1 | TC0X00010056.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | ENST00000438453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010056.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168639.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168639.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168639.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168639.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168639.hg.1 | TC0X00010056.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00168642.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168642.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168642.hg.1 | ENST00000602584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168642.hg.1 | NR_029423.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168642.hg.1 | TC0X00010056.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040072 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040073 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040073 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A040072 | - | - |

MAP2K4P1
Genomic Map