Assay Details
| Gene Symbol: RBAK-RBAKDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204513.1 | NP_001191442.1 | 4 - 5 | 422 | 57 | |
| GenBank mRNA | BM684776.1 | - | 2 - 3 | 57 | ||
| BM929719.1 | - | 4 - 5 | 332 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003318.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07000780.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000780.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000780.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000780.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000780.hg.1 | TC07003318.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | ENST00000353796.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000765.hg.1 | TC07003318.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000763.hg.1 | ENST00000353796.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000763.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000763.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000763.hg.1 | TC07003318.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000476992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013332.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196258.hg.1 | TC0700013332.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | ENST00000498308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | NR_015343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196277.hg.1 | TC0700013332.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | ENST00000353796.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | ENST00000396904.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | ENST00000396912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | NM_001204456.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | NM_001204513.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | NM_021163.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196260.hg.1 | TC0700013332.hg.1 | Clariom D TC |

RBAK-RBAKDN
Genomic Map