Assay Details
| Gene Symbol: MLLT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195628.1 | NP_001182557.1 | 1 - 2 | 329 | 64 | |
| NR_136736.1 | 1 - 2 | 329 | 64 | |||
| GenBank mRNA | AB209755.1 | - | 1 - 2 | 35 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195628.1 transcripts.
Set Membership: |
|

MLLT10
Genomic Map