Assay Details
| Gene Symbol: WISP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204869.1 | NP_001191798.1 | 2 - 3 | 457 | 68 | |
| GenBank mRNA | AY196488.1 | - | 2 - 3 | 412 | 68 | |
More Information
Set Membership: |
|

WISP1
Genomic Map