Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001745.3 | NP_001736.1 | 2 - 3 | 778 | 78 | |
| GenBank mRNA | AF068179.1 | - | 2 - 3 | 495 | 78 | |
| AK313156.1 | - | 2 - 3 | 702 | 78 | ||
| BC130325.1 | - | 2 - 3 | 710 | 40146356 | 78 | |
| U18242.1 | - | 2 - 3 | 673 | 78 | ||
More Information
Set Membership: |
|

CAMLG
Genomic Map