Assay Details
| Gene Symbol: COX6A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005205.3 | NP_005196.1 | 1 - 2 | 180 | 72 | |
| GenBank mRNA | BC029818.1 | - | 1 - 2 | 140 | 5174639 | 72 |
| BP319904.1 | - | 180 | 72 | |||
| F27891.1 | - | 1 - 2 | 143 | 72 | ||
| HE647858.1 | - | 1 - 2 | 77 | 72 | ||
| M83308.1 | - | 1 - 2 | 113 | 72 | ||
More Information
Set Membership: |
|

COX6A2
Genomic Map