Assay Details
| Gene Symbol: MIR615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030753.1 | 1 | 49 | 54 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox C6 | HOXC6 |
CP25, HHO.C8, HOX3, HOX3C |
-3.1kb | Sense | Chr. 12 - 54016852 - 54030823 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC440101 | FLJ12825 | 24.2kb | Sense | Chr. 12 - 54058254 - 54122234 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1200007742.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000453.hg.1
Set Membership: |
|

MIR615
Genomic Map