Assay Details
| Gene Symbol: MIR96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029512.1 | 1 | 39 | 64 | ||
| GenBank mRNA | AF480568.1 | - | 1 | 39 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR96| Gene Symbol: MIR96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029512.1 | 1 | 39 | 64 | ||
| GenBank mRNA | AF480568.1 | - | 1 | 39 | 64 | |
Set Membership: |
|