Assay Details
| Gene Symbol: CLSTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009566.2 | NP_001009566.1 | 2 - 3 | 1008 | 114 | |
| NM_001302883.1 | NP_001289812.1 | 2 - 3 | 1008 | 114 | ||
| GenBank mRNA | AB020718.1 | - | 2 - 3 | 1008 | 114 | |
| AK304813.1 | - | 2 - 3 | 250 | 114 | ||
More Information
Set Membership: |
|

CLSTN1
Genomic Map