Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK223393.1 | - | 3 | 438 | 69 | |
| AK301154.1 | - | 3 | 461 | 69 | ||
| AK303367.1 | - | 3 | 434 | 69 | ||
| AK313614.1 | - | 3 | 467 | 69 | ||
| BC033583.1 | - | 3 | 422 | 5227138 | 69 | |
| CD691699.1 | - | 3 | 492 | 69 | ||
| CR996443.1 | - | 3 | 444 | 69 | ||
| M14362.1 | - | 3 | 418 | 69 | ||
| M16336.1 | - | 3 | 432 | 69 | ||
| M16445.1 | - | 3 | 415 | 69 | ||
More Information
Set Membership: |
|

CD2
Genomic Map