Assay Details
| Gene Symbol: MIR365A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029854.1 | 1 | 40 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 193b | MIR193B |
MIRN193B, mir-193b |
-5.2kb | Sense | Chr. 16 - 14303967 - 14304049 on NCBI Build GRCh38 |
| Downstream | MIR193B host gene | MIR193BHG | 12.1kb | Sense | Chr. 16 - 14302288 - 14326517 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1600006988.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000175.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451704 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939245 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30451704 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16853258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09939222 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7993337 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002633.hg.1 | ENST00000362260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002633.hg.1 | NR_029854.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002633.hg.1 | TC16000175.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006988.hg.1 | ENST00000362260.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006988.hg.1 | NR_029854.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006988.hg.1 | ENST00000362260.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006988.hg.1 | NR_029854.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149533.hg.1 | ENST00000362260.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149533.hg.1 | NR_029854.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149533.hg.1 | TC1600006988.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600149533.hg.1 | ENST00000362260.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149533.hg.1 | NR_029854.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309443 | NR_029854.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309443 | NR_029854.1 | human_transcriptome |

MIR365A
Genomic Map