Assay Details
| Gene Symbol: MIR578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030304.1 | 1 | 45 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methylsterol monooxygenase 1 | MSMO1 |
DESP4, ERG25, MCCPD, SC4MOL |
-43.1kb | Sense | Chr. 4 - 165327666 - 165343164 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1179 | LINC01179 | 298.3kb | Sense | Chr. 4 - 165684639 - 165762778 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0400009224.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000830.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15635146 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592452 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754292 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592469 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15635133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592475 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15635150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592475 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592452 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15635146 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592469 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754290 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15635133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15635150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754292 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754318 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23754293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03592449 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009224.hg.1 | ENST00000384828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009224.hg.1 | NR_030304.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011474.hg.1 | ENST00000384828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011474.hg.1 | NR_030304.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011474.hg.1 | TC04000830.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400163404.hg.1 | ENST00000384828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163404.hg.1 | NR_030304.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400163404.hg.1 | ENST00000384828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400163404.hg.1 | NR_030304.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400163404.hg.1 | TC0400009224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009224.hg.1 | ENST00000384828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009224.hg.1 | NR_030304.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310995 | NR_030304.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310995 | NR_030304.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310995 | NR_030304.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310995 | NR_030304.1 | human_transcriptome |

MIR578
Genomic Map