| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002307.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002307.hg.1 |
ENST00000385082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002307.hg.1 |
NR_030345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12002307.hg.1 |
TC12000182.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000385082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000534831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000534831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200006896.hg.1 |
NR_030345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000385082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000534831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000534831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000537783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000540125.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
ENST00000542056.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NM_003979.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200006896.hg.1 |
NR_030345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
ENST00000385082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
NR_030345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
ENST00000014914.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
ENST00000385082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
NR_030345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200148220.hg.1 |
TC1200006896.hg.1 |
Clariom D TC |