Assay Details
| Gene Symbol: MIR320C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031565.1 | 1 | 48 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | abhydrolase domain containing 3 | ABHD3 | LABH3 | Anti-sense | Chr. 18 - 21650897 - 21704805 on NCBI Build GRCh38 | |
| Downstream | mindbomb E3 ubiquitin protein ligase 1 | MIB1 |
DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6 |
57.2kb | Sense | Chr. 18 - 21740793 - 21870957 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1800006840.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000089.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Chr array_18 19 21 | A_17_P10563554 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048582 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563560 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162201 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162210 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563549 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162224 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162221 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162226 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162195 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162206 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563571 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162217 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563555 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563568 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162223 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563547 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162194 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162213 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162212 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048579 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563576 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048581 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162209 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162204 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563550 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563551 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563552 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563567 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563557 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563548 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563558 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563575 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563569 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563565 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048580 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563564 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048584 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563567 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563564 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563550 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563552 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563549 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563550 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162226 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563557 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162204 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162217 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563571 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563555 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563567 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048582 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162213 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048584 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162210 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563564 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563568 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048581 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048580 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162195 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162194 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563548 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162209 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162223 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563576 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563547 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162212 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563551 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563569 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563560 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162206 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563575 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162221 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563552 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563554 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162224 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563558 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162208 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10563565 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31162201 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17048579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563550 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563555 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563567 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563552 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17048579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10563564 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31162204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8020419 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001007.hg.1 | ENST00000408566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001007.hg.1 | NR_031565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001007.hg.1 | TC18000089.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011000.hg.1 | ENST00000390179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011000.hg.1 | ENST00000390179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011000.hg.1 | NR_036157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006840.hg.1 | ENST00000408566.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006840.hg.1 | NR_031565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146844.hg.1 | ENST00000408566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146844.hg.1 | NR_031565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146844.hg.1 | TC1800006840.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146844.hg.1 | ENST00000408566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146844.hg.1 | NR_031565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006840.hg.1 | ENST00000408566.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006840.hg.1 | NR_031565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011000.hg.1 | ENST00000390179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011000.hg.1 | ENST00000390179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011000.hg.1 | NR_036157.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3311125 | NR_031565.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3311125 | NR_031565.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3311125 | NR_031565.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3311125 | NR_031565.1 | human_transcriptome |

MIR320C1
Genomic Map