Assay Details
| Gene Symbol: MIR764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031581.1 | 1 | 42 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 35 |
SNORA35 |
HBI-36, SNORA35A |
-8.5kb | Sense | Chr. X - 114630797 - 114630924 on NCBI Build GRCh38 |
| Downstream | microRNA 1912 | MIR1912 |
MIRN1912, hsa-mir-1912, mir-1912 |
12.0kb | Sense | Chr. X - 114651544 - 114651623 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00008182.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000557.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17388743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899096 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629143 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629156 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629144 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899096 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899086 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629133 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629158 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629149 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629144 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629116 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629143 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17388743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629156 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32629129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11899145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008182.hg.1 | ENST00000390811.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008182.hg.1 | NR_031581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007428.hg.1 | ENST00000390811.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007428.hg.1 | NR_031581.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007428.hg.1 | TC0X000557.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00008182.hg.1 | ENST00000390811.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008182.hg.1 | NR_031581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155834.hg.1 | ENST00000390811.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155834.hg.1 | NR_031581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155834.hg.1 | ENST00000390811.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155834.hg.1 | NR_031581.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155834.hg.1 | TC0X00008182.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308240 | NR_031581.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308240 | NR_031581.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308240 | NR_031581.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308240 | NR_031581.1 | human_transcriptome |

MIR764
Genomic Map