Assay Details
| Gene Symbol: MIR1179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031590.1 | 1 | 47 | 91 | ||
| Gene Symbol: AEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017022489.1 | XP_016877978.1 | 1 | 375 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1586 | LINC01586 | -3.0kb | Anti-sense | Chr. 15 - 88585571 - 88605110 on NCBI Build GRCh38 | |
| Downstream | microRNA 7-2 | MIR7-2 |
MIRN7-2, hsa-mir-7-2, mir-7-2 |
3.6kb | Sense | Chr. 15 - 88611825 - 88611934 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1500008229.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15000838.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017022489.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816915 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816931 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816917 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816930 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816910 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816947 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816930 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816915 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816931 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816933 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816923 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816910 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816947 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816937 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816903 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816917 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816928 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09816891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30320818 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15007711.hg.1 | ENST00000408703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007711.hg.1 | NR_031590.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007711.hg.1 | TC15000838.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008229.hg.1 | ENST00000408703.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008229.hg.1 | NR_031590.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008229.hg.1 | ENST00000408703.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008229.hg.1 | NR_031590.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500159257.hg.1 | ENST00000408703.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500159257.hg.1 | NR_031590.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500159257.hg.1 | TC1500008229.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500159257.hg.1 | ENST00000408703.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500159257.hg.1 | NR_031590.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308205 | NR_031590.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308205 | NR_031590.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308205 | NR_031590.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308205 | NR_031590.1 | human_transcriptome |

MIR1179
Genomic Map