Assay Details
| Gene Symbol: MIR1284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031697.1 | 1 | 63 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FOXP1 antisense RNA 1 | FOXP1-AS1 | -236.1kb | Anti-sense | Chr. 3 - 71289758 - 71305854 on NCBI Build GRCh38 | |
| Downstream | eukaryotic translation initiation factor 4E family member 3 | EIF4E3 |
eIF-4E3, eIF4E-3 |
137.2kb | Sense | Chr. 3 - 71679289 - 71754831 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300011512.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001547.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332915 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332951 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15450766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332884 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332938 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332936 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332936 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332915 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468182 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332921 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332951 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332938 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332884 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468198 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15450766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468175 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02332887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22468205 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03028237.hg.1 | ENST00000408337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028237.hg.1 | NR_031697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028237.hg.1 | TC03001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011512.hg.1 | ENST00000408337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011512.hg.1 | NR_031697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011512.hg.1 | ENST00000408337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011512.hg.1 | NR_031697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186160.hg.1 | ENST00000408337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186160.hg.1 | NR_031697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186160.hg.1 | ENST00000408337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300186160.hg.1 | NR_031697.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300186160.hg.1 | TC0300011512.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309499 | NR_031697.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309499 | NR_031697.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309499 | NR_031697.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309499 | NR_031697.1 | human_transcriptome |

MIR1284
Genomic Map