| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019544.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019544.hg.1 |
TC03001140.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019545.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019545.hg.1 |
TC03001140.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019546.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019546.hg.1 |
TC03001140.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019547.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019547.hg.1 |
TC03001140.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019542.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019542.hg.1 |
TC03001140.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206338.hg.1 |
TC0300013928.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206340.hg.1 |
TC0300013928.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206341.hg.1 |
TC0300013928.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000420095.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000452802.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
ENST00000455811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013928.hg.1 |
NR_033378.1 |
Refseq |