| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009407.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013843.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013843.hg.1 |
TC14001213.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013844.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013844.hg.1 |
TC14001213.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400169481.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169481.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169481.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169481.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169481.hg.1 |
TC1400009407.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
ENST00000556556.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009407.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169477.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169477.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169477.hg.1 |
TC1400009407.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400169478.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169478.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169478.hg.1 |
ENST00000447107.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169478.hg.1 |
NR_033777.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169478.hg.1 |
TC1400009407.hg.1 |
Clariom D TC |