| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000048.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000048.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000048.hg.1 |
TC07000007.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000049.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000049.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000049.hg.1 |
TC07000007.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000050.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000050.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07000050.hg.1 |
TC07000007.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700144210.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144210.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144210.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144210.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144210.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144223.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144223.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144223.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144223.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144223.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006458.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144211.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144211.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144211.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144211.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144211.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144216.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144216.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144216.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144216.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144216.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144207.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144207.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144207.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144207.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144207.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144209.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144209.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144209.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144209.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144209.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144221.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144221.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144221.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144221.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144221.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144222.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144222.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144222.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144222.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144222.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144208.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144208.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144208.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144208.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144208.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700144212.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144212.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144212.hg.1 |
ENST00000452622.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700144212.hg.1 |
NR_033963.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700144212.hg.1 |
TC0700006458.hg.1 |
Clariom D TC |