| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044489.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044489.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044489.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044489.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044489.hg.1 |
TC01002851.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044487.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044487.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044487.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044487.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044487.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044487.hg.1 |
TC01002851.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044491.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044491.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01044491.hg.1 |
TC01002851.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210785.hg.1 |
TC0100014864.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210786.hg.1 |
TC0100014864.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014864.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210783.hg.1 |
TC0100014864.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000415584.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000443562.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000526694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
ENST00000528692.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
NR_033981.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210779.hg.1 |
TC0100014864.hg.1 |
Clariom D TC |