Assay Details
| Gene Symbol: FLJ27354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033981.1 | 2 - 3 | 428 | 89 | ||
| GenBank mRNA | AK130864.1 | - | 2 - 3 | 428 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 8 family member C | LRRC8C |
AD158, FAD158 |
Anti-sense | Chr. 1 - 89615810 - 89719535 on NCBI Build GRCh38 | |
| Downstream | leucine rich repeat containing 8 family member D | LRRC8D | LRRC5 | 188.8kb | Anti-sense | Chr. 1 - 89821014 - 89936432 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002851.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395349 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395372 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395369 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00365832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395348 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20395362 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | AK025682.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | AK074260.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | AK127061.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | AK225323.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | AL832424.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | BC053873.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | BX647378.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | BX647704.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 102090 | NM_024949.5 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 894414 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 169030 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 667541 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 2513 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 59420 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15209046 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00118549 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10220476 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35218200 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15209063 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35218200 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15209046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 231313_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000526694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000526694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000526694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000528692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000528692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | ENST00000528692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044484.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044484.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044484.hg.1 | TC01002851.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000443562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000526694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000526694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000526694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000528692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000528692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | ENST00000528692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014864.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210769.hg.1 | TC0100014864.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | ENST00000415584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | NR_033981.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210770.hg.1 | TC0100014864.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 212978_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 212978_at | - | - |
| Human Genome U133 Plus 2.0 Array | 231313_at | - | - |
| Human Genome U133 Plus 2.0 Array | 231313_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231313_at5 | - | - |
| Human Genome U95 Set | 76234_PM_at | - | - |
| Human Genome U95 Set | 76234_at | - | - |
| Human Genome U95 Set | 73774_PM_at | - | - |
| Human Genome U95 Set | 73774_at | - | - |
| Human Genome U133 Set | 231313_at | - | - |
| Human Genome U133 Set | 231313_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048034 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048034 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 212871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1720004 | NR_033981.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1720004 | AK130864.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720004 | NR_033981.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1720004 | AK130864.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720004 | NR_033981.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1720004 | AK130864.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720004 | NR_033981.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1720004 | AK130864.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720004 | NR_033981.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1720004 | AK130864.1 | human_transcriptome |

FLJ27354
Genomic Map