Assay Details
| Gene Symbol: IGFBP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034081.1 | 1 - 2 | 210 | 114 | ||
| GenBank mRNA | BC039519.1 | - | 1 - 2 | 210 | 114 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM546357.1 | - | 229 | Chr. - - [-] on Build | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA polymerase II subunit B | POLR2B |
POL2RB, RPB2, hRPB140 |
-78.6kb | Sense | Chr. 4 - 56978640 - 57031162 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928851 | LOC101928851 | 237.6kb | Sense | Chr. 4 - 57425872 - 57465986 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000339.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04004822.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004822.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004822.hg.1 | TC04000339.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004825.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004825.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004825.hg.1 | TC04000339.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004823.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004823.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004823.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004823.hg.1 | TC04000339.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152142.hg.1 | TC0400007621.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152158.hg.1 | TC0400007621.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000511330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | ENST00000515865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007621.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152157.hg.1 | TC0400007621.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152140.hg.1 | TC0400007621.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000499667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | ENST00000508328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | NR_034081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400152141.hg.1 | TC0400007621.hg.1 | Clariom D TC |

IGFBP7-AS1
Genomic Map