Assay Details
| Gene Symbol: LINC01160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034126.1 | 2 - 3 | 258 | 132 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane and immunoglobulin domain containing 3 | TMIGD3 | AD026 | -40.4kb | Sense | Chr. 1 - 111483348 - 111564003 on NCBI Build GRCh38 |
| Downstream |
RAP1A, member of RAS oncogene family |
RAP1A |
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21 |
15.1kb | Anti-sense | Chr. 1 - 111542223 - 111716695 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002988.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01046407.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046407.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046407.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046407.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046407.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046409.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046409.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046409.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046409.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046408.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046408.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046408.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046408.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046408.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046410.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046410.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046410.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046410.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046410.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213954.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213956.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213955.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213957.hg.1 | TC0100015280.hg.1 | Clariom D TC |

LINC01160
Genomic Map