Assay Details
| Gene Symbol: LINC01160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034126.1 | 3 - 4 | 390 | 98 | ||
| GenBank mRNA | AL553535.3 | - | 3 - 4 | 391 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane and immunoglobulin domain containing 3 | TMIGD3 | AD026 | -38.2kb | Sense | Chr. 1 - 111483348 - 111564003 on NCBI Build GRCh38 |
| Downstream |
RAP1A, member of RAS oncogene family |
RAP1A |
C21KG, G-22K, KREV-1, KREV1, RAP1, SMGP21 |
15.3kb | Anti-sense | Chr. 1 - 111542223 - 111716695 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002988.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459053 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491733 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491713 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491749 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491748 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491694 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491706 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20491700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15099942 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00459055 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3399768 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046402.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046402.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046402.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046403.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046403.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046403.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046403.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046404.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046404.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046404.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046404.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046401.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046401.hg.1 | TC01002988.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | ENST00000453988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015280.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213951.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213949.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000441004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | ENST00000450155.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213950.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100213948.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213948.hg.1 | NR_034126.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100213948.hg.1 | TC0100015280.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A000319 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A000319 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3260111 | NR_034126.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3260111 | CR620722.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3180420 | NR_034126.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3180420 | CR620722.1 | human_transcriptome |

LINC01160
Genomic Map