| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012145.hg.1 |
ENST00000583685.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012145.hg.1 |
NR_036092.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022989.hg.1 |
NR_036092.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04022989.hg.1 |
TC04001648.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400012145.hg.1 |
ENST00000583685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012145.hg.1 |
NR_036092.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183413.hg.1 |
ENST00000583685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183413.hg.1 |
NR_036092.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183413.hg.1 |
ENST00000583685.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183413.hg.1 |
NR_036092.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183413.hg.1 |
TC0400012145.hg.1 |
Clariom D TC |