Assay Details
| Gene Symbol: MIR3915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037478.1 | 1 | 51 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ferritin heavy chain like 17 | FTHL17 | CT38 | -1.5mb | Sense | Chr. X - 31071241 - 31072053 on NCBI Build GRCh38 |
| Downstream | microRNA 548f-5 | MIR548F5 |
MIR548F-5, MIRN548F5, hsa-mir-548f-5 |
57.7kb | Sense | Chr. X - 32641474 - 32641559 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00009343.hg.1
Set Membership: |
|

MIR3915
Genomic Map