| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002507.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002507.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002507.hg.1 |
TC04000159.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002508.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002508.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002508.hg.1 |
TC04000159.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002509.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002509.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04002509.hg.1 |
TC04000159.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400148180.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400148180.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148180.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400148180.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148180.hg.1 |
TC0400007018.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400148179.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400148179.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148179.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400148179.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148179.hg.1 |
TC0400007018.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007018.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148181.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400148181.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148181.hg.1 |
ENST00000515882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400148181.hg.1 |
NR_037876.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400148181.hg.1 |
TC0400007018.hg.1 |
Clariom D TC |