| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000428561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000428561.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006888.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006888.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006888.hg.1 |
TC03003356.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006887.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006887.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006887.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03006887.hg.1 |
TC03003356.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204524.hg.1 |
TC0300013836.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204523.hg.1 |
TC0300013836.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300204525.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204525.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204525.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300204525.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300204525.hg.1 |
TC0300013836.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000414493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000422896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000428561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000428561.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000433169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
ENST00000441610.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038213.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038214.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013836.hg.1 |
NR_038215.1 |
Refseq |
| Human Genome U95 Set |
55527_at |
- |
- |
| Human Genome U95 Set |
55529_g_at |
- |
- |
| Human Genome U95 Set |
55527_PM_at |
- |
- |
| Human Genome U95 Set |
55529_PM_g_at |
- |
- |