| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023992.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023992.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023992.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023992.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12023992.hg.1 |
TC12001817.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011541.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000499137.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000500986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000547845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
ENST00000551626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
NR_038263.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189443.hg.1 |
TC1200011541.hg.1 |
Clariom D TC |