Assay Details
| Gene Symbol: MYLK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038266.2 | 1 - 2 | 290 | 100 | ||
| NR_121654.1 | 2 - 3 | 197 | 100 | |||
| GenBank mRNA | BC042384.1 | - | 1 - 2 | 246 | 100 | |
| DA531919.1 | - | 1 - 2 | 273 | 100 | ||
| DA714112.1 | - | 2 - 3 | 202 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 3-hydroxyacyl-CoA dehydratase 2 | HACD2 | PTPLB | -436.0 | Anti-sense | Chr. 3 - 123491548 - 123585093 on NCBI Build GRCh38 |
| Downstream | MYLK antisense RNA 2 | MYLK-AS2 | 60.0kb | Sense | Chr. 3 - 123689644 - 123692407 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000639.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690976 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553172 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15481738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690960 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02553166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22690965 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 109332 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 62202 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 336495 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 179376 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 568994 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 499241 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 138620 | AK024854.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 138620 | AY743217.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 138620 | NM_004900.3 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 138620 | U61083.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 138620 | U61084.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 489927 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 215267 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16387753 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16387734 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P16387753 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16387738 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16387775 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16387765 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16387734 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16387753 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 116186 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 77610 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00801793 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387738 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387735 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387756 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387773 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36432462 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387765 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36432460 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36432482 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387753 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387734 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P36432498 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387781 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16387775 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16387753 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16387734 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16387738 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16387775 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16387765 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03011553.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011553.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011553.hg.1 | TC03000639.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011558.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011558.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011558.hg.1 | TC03000639.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011557.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011557.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011557.hg.1 | TC03000639.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011555.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011555.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011555.hg.1 | TC03000639.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161135.hg.1 | TC0300008582.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161120.hg.1 | TC0300008582.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161130.hg.1 | TC0300008582.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008582.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161117.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161117.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161117.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161117.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161117.hg.1 | TC0300008582.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161134.hg.1 | TC0300008582.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000463408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000470449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | ENST00000485162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | NR_038266.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | NR_121654.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161124.hg.1 | TC0300008582.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023105 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023105 | - | - |

MYLK-AS1
Genomic Map