| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019070.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019070.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019070.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019070.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019070.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019070.hg.1 |
TC03001105.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300173161.hg.1 |
TC0300010046.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
ENST00000446695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010046.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000414354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000415244.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000424769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
ENST00000437064.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
NR_038285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300173162.hg.1 |
TC0300010046.hg.1 |
Clariom D TC |