Assay Details
| Gene Symbol: MAGI2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038346.1 | 2 - 3 | 302 | 128 | ||
| GenBank mRNA | BI868593.1 | - | 2 - 3 | 101 | 128 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAGI2 antisense RNA 2 | MAGI2-AS2 | -441.3kb | Sense | Chr. 7 - 79008988 - 79012277 on NCBI Build GRCh38 | |
| Downstream | G protein subunit alpha i1 | GNAI1 | Gi | 675.8kb | Sense | Chr. 7 - 80134824 - 80219409 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MAGI2-AS3
Genomic Map