| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006758.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006758.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006758.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006758.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001703.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001703.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001703.hg.1 |
TC01000108.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001704.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001704.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001704.hg.1 |
TC01000108.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100146748.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146748.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146748.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146748.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146748.hg.1 |
TC0100006758.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100006758.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006758.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006758.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006758.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146747.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146747.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146747.hg.1 |
ENST00000442636.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100146747.hg.1 |
NR_038351.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100146747.hg.1 |
TC0100006758.hg.1 |
Clariom D TC |