| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000555801.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000555801.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
ENST00000557295.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003103.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003103.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003103.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003103.hg.1 |
TC14000296.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003099.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003099.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003099.hg.1 |
TC14000296.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003104.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003104.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003104.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003104.hg.1 |
TC14000296.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149478.hg.1 |
TC1400007142.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000555801.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000555801.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
ENST00000557295.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007142.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
ENST00000554409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149477.hg.1 |
TC1400007142.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
ENST00000556479.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
NR_038358.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400149473.hg.1 |
TC1400007142.hg.1 |
Clariom D TC |