Assay Details
| Gene Symbol: LINC00640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038358.1 | 3 - 4 | 302 | 129 | ||
| GenBank mRNA | BX111479.1 | - | 3 - 4 | 154 | 129 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thioredoxin related transmembrane protein 1 | TMX1 |
PDIA11, TMX, TXNDC, TXNDC1 |
-92.0kb | Sense | Chr. 14 - 51240168 - 51257655 on NCBI Build GRCh38 |
| Downstream | FRMD6 antisense RNA 2 | FRMD6-AS2 | 104.1kb | Anti-sense | Chr. 14 - 51454512 - 51599952 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000296.hg.1
Set Membership: |
|

LINC00640
Genomic Map