Assay Details
| Gene Symbol: LOC644838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038844.1 | 2 - 3 | 140 | 83 | ||
| GenBank mRNA | AK123766.1 | - | 2 - 3 | 140 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102800447 | LOC102800447 | Anti-sense | Chr. 2 - 67086446 - 67289244 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC101927661 | LOC101927661 | 7.8kb | Anti-sense | Chr. 2 - 67213408 - 67244574 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001935.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02030720.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030720.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030720.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030720.hg.1 | TC02001935.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030723.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030723.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030723.hg.1 | TC02001935.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030715.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030715.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030715.hg.1 | TC02001935.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030721.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030721.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030721.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030721.hg.1 | TC02001935.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030716.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030716.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030716.hg.1 | TC02001935.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030722.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030722.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030722.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02030722.hg.1 | TC02001935.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193901.hg.1 | TC0200012908.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193913.hg.1 | TC0200012908.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193908.hg.1 | TC0200012908.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193912.hg.1 | TC0200012908.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | ENST00000414404.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193909.hg.1 | TC0200012908.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | ENST00000433133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | NR_038844.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200193902.hg.1 | TC0200012908.hg.1 | Clariom D TC |

LOC644838
Genomic Map