| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030713.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030713.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030713.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030713.hg.1 |
TC02001935.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030714.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030714.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02030714.hg.1 |
TC02001935.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193899.hg.1 |
TC0200012908.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000414404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
ENST00000447453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012908.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
ENST00000433133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
NR_038844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193900.hg.1 |
TC0200012908.hg.1 |
Clariom D TC |