Assay Details
| Gene Symbol: LOC284950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038888.1 | 1 - 2 | 228 | 66 | ||
| GenBank mRNA | AK095038.1 | - | 1 - 2 | 229 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | atonal bHLH transcription factor 8 | ATOH8 |
HATH6, bHLHa21 |
-23.7kb | Sense | Chr. 2 - 85753786 - 85791383 on NCBI Build GRCh38 |
| Downstream | ST3 beta-galactoside alpha-2,3-sialyltransferase 5 | ST3GAL5 |
SATI, SIAT9, SIATGM3S, ST3GalV |
20.8kb | Anti-sense | Chr. 2 - 85839148 - 85889034 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000518.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331920 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15263863 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331871 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414539 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331928 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15263867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15263843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414545 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331930 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331885 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414562 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15263864 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331880 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21414514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331890 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331937 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15263859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331900 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01331864 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 294402 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 515223 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 835863 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 509681 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3791552 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26121 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3791552 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3791552 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15733272 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 97140 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13293976 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15733268 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00421645 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15733272 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00421644 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00421645 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15733272 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563680_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008046.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008046.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008046.hg.1 | TC02000518.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008047.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008047.hg.1 | TC02000518.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008044.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008044.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008044.hg.1 | TC02000518.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008045.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008045.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008045.hg.1 | TC02000518.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200157408.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157408.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157408.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008271.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157406.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157406.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157406.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157404.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157404.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157404.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157404.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157404.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157407.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157407.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157407.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157401.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157401.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157401.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157401.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157401.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157403.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157403.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157403.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157403.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157403.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157405.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157405.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157405.hg.1 | ENST00000366278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157405.hg.1 | NR_038888.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157405.hg.1 | TC0200008271.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1563680_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563680_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563680_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015917 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015917 | - | - |

LOC284950
Genomic Map