Assay Details
| Gene Symbol: DLGAP1-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038895.1 | 1 - 2 | 114 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371967 | LOC105371967 | -221.9kb | Sense | Chr. 18 - 3653056 - 3656282 on NCBI Build GRCh38 | |
| Downstream | DLGAP1 antisense RNA 4 | DLGAP1-AS4 | 66.4kb | Sense | Chr. 18 - 3962353 - 4013943 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000023.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514911 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105573 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514927 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514864 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033282 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105595 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033293 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105605 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105628 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105647 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105631 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105644 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105657 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105575 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514888 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105658 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105559 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105604 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105572 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105588 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105592 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105566 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105629 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17033273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105654 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10514893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31105642 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3511777 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36160 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3511777 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3511777 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20758440 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P40934793 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 157213 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40934824 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03307127 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12768899 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P21018014 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20758440 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20758424 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12769975 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P34287765 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40934793 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1560522_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000245.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000245.hg.1 | TC18000023.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000247.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000247.hg.1 | TC18000023.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000246.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000246.hg.1 | TC18000023.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | TC1800006528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144740.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | TC1800006528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144739.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144736.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144736.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144736.hg.1 | TC1800006528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144736.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144736.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144738.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144738.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144738.hg.1 | TC1800006528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144738.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144738.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | ENST00000583571.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006528.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144741.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144741.hg.1 | NR_038895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144741.hg.1 | TC1800006528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144741.hg.1 | ENST00000577649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144741.hg.1 | NR_038895.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1560522_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560522_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560522_at3 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010449 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010449 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 132378 | - | - |

DLGAP1-AS3
Genomic Map